missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde, 99%

Catalog No. AAA1383336
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1383336 500 g
AAA1383322 100 g
AAA138330E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1383336 Supplier Thermo Scientific Chemicals Supplier No. A1383336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 90-02-8 | C7H6O2 | 122.12 g/mol

Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.

Solubility
Miscible with alcohol, ether, acetone, benzene and most organic solvents. Slightly miscible with water.

Notes
Incompatible with acids, strong reducing agents, strong oxidizing agents, strong bases and halogens.

Chemical Identifiers

CAS 90-02-8
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003317
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
Synonym salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
PubChem CID 6998
ChEBI CHEBI:16008
IUPAC Name 2-hydroxybenzaldehyde
SMILES OC1=CC=CC=C1C=O

Specifications

Melting Point °C to 7°C
Density 1.168
Boiling Point 197°C
Flash Point 76°C (168°F)
Assay Percent Range 99%
Refractive Index 1.573
Quantity 500 g
Beilstein 471388
Sensitivity Air and light sensitive
Merck Index 14,8326
Solubility Information Miscible with alcohol,ether,acetone,benzene and most organic solvents. Slightly miscible with water.
Formula Weight 122.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Salicylaldehyde
Show More Show Less
Hazard Category H227-H302-H311-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P309+P311-P312-P314-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 201-961-0
RTECSNumber VN5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only