missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde, 98%, Spectrum™ Chemical

Catalog No. 18606142
Change view
Click to view available options
Quantity:
1 L
200 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606142 200 L Amber Glass Bottle
18606141 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18606142 Supplier Spectrum Chemical Mfg Cor Supplier No. SA122200LTBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SA122, 90-02-8, C7H6O2

Spectrum™ Chemical Salicylaldehyde , also known as Salicylic aldehyde or 2-Hydroxybenzaldehyde, is a precursor to a several types of commercially important chelating agents. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 90-02-8
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003317
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
IUPAC Name 2-hydroxybenzaldehyde
SMILES OC1=CC=CC=C1C=O

Specifications

CAS Max % 1
Boiling Point 197°C (386.6°F)
Packaging Amber Glass Bottle
Refractive Index 1.571-1.575
Quantity 200 L
Percent Purity 98%
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Salicylaldehyde