missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldazine, 99%

Catalog No. AC456941000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456941000 100 g
AC456940050 5 g
AC456940250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC456941000 Supplier Thermo Scientific Chemicals Supplier No. 456941000

CAS: 959-36-4 | C14H12N2O2 | 240.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 959-36-4
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
InChI Key SPEXYYIULCBQJR-UHFFFAOYSA-N
Synonym salicylaldehyde azine, salicylalazine, salicylaldazine, benzaldehyde, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazone, 2-hydroxybenzaldehyde azine, 2-1e-e-2-2-hydroxyphenyl methylidene hydrazin-1-ylidene methyl phenol, 2,2'-1,2-hydrazinediylidenedi methylylidene diphenol, benzaldehyde, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazone, 1,2-bis e-salicylidene hydrazine, 6-6-oxo-1-cyclohexa-2,4-dienylidene methylhydrazo methylidene-1-cyclohexa-2,4-dienone
PubChem CID 6849893
IUPAC Name 6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one
SMILES C1=CC(=CNNC=C2C=CC=CC2=O)C(=O)C=C1

Specifications

Melting Point 218.0°C to 220.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 100 g
Solubility Information Solubility in water: insoluble.
Formula Weight 240.26
Percent Purity 99%
Chemical Name or Material Salicylaldazine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
EINECSNumber 213-500-

RUO – Research Use Only