missing translation for 'onlineSavingsMsg'
Learn More

Salcomine 95.0+%, TCI America™

Catalog No. S031825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031825G 25 g
S0318100G 100 g
S0318500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S031825G Supplier TCI America Supplier No. S031825G

Chemical Identifiers

CAS 14167-18-1
Molecular Formula C16H14CoN2O2
Molecular Weight (g/mol) 325.23
MDL Number MFCD00000009
InChI Key NPAQSKHBTMUERN-UHFFFAOYSA-L
PubChem CID 57448789
IUPAC Name λ²-cobalt(2+) 2-{[(2-{[(2-oxidophenyl)methylidene]amino}ethyl)imino]methyl}benzen-1-olate
SMILES [Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]

Specifications

Quantity 25 g
Synonym N,N′C-Bis(salicylidene)ethylenediiminocobalt(II), N,N′C-Disalicylalethylenediamine Cobalt(II), N,N′C-Ethylenebis(salicylideneiminato)cobalt(II)
Formula Weight 325.23
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Salcomine [=N,N′-Bis(salicylidene)ethylenediiminocobalt(II)]
RTECSNumber GG0590000
TSCA Yes