missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salbutamol Aldehyde Impurity, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 156339-88-7 |
| Molecular Formula | C13H19NO3 |
| Linear Formula | C13H19NO3 |
| Synonym | 5-(2-tert-Butylamino-1-hydroxyethyl)-2-hydroxybenzaldehyde; 5-[(1 RS)-2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-2-hydroxybenzaldehyde |
| Type | Pharmacopeia and Metrological Institutes Standards |
| Quantity | 25 mg |
| Formula Weight | 237.29 |
| Grade | British Pharmacopoeia (BP) Reference Standard |