missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Saint-Gobain™ Tygon™ Sanitary Silicone Tubing Formula 3350

Translucent tubing ideal for biotechnological, pharmaceutical, and peristaltic applications

Manufacturer:  Saint-Gobain™ ABW00053

 View more versions of this product

Catalog No. 02-587-2F



Description

Description

Translucent tubing ideal for biotechnological, pharmaceutical, and peristaltic applications

Ultrasmooth inner bore reduces risk of particle entrapment, inhibits microscopic bacterial buildup during fluid transfer, and allows thorough cleaning and sterilization.

  • Tasteless, odorless
  • Nonpyrogenic, nontoxic, nonhemolytic
  • Low extractable content helps to maintain integrity and purity of transported fluids
  • Smooth inner surface provides low adsorption for improved fluid flow (hydrophobic surface also improves fluid flow)
  • Maximum recommended operating temperature 200°C (392°F)
  • Autoclavable; also can be radiation- and gas-sterilized
  • Durometer hardness: Shore A, 50

Not intended for use as an implant material.

Order Info

Packaged in heat-sealed, double-wrapped polyethylene packs of 50 ft. (15.2cm)

Compliance

Complies with requirements of USP XXII Class VI criteria. Meets 3A Sanitary Standards, FDA 21 CFR Part 177.2600 criteria and NSF 51 Standard.

Specifications

Specifications

Complies with requirements of USP XXII Class VI criteria. Meets 3A Sanitary Standards, FDA 21 CFR Part 177.2600 criteria and NSF 51 Standard.
3/4 in.
Tygon
25.4mm
3.2mm
Yes
19mm
1 in.
0.125 in.
Documents
Accessories