Learn More
Saint-Gobain Tygon S3™ Flexible Tubing, Formula B-44-4X I.B.
For food, beverage, dairy, and other bacterial-sensitive applications
Supplier:  Saint-Gobain AMH00064
Encompass
Catalog No. 14171110
Description
Description
For food, beverage, dairy, and other bacterial-sensitive applications
  • Handles four times pressure of non-reinforced tubing
  • Smooth, non-porous bore resists particle entrapment and resultant bacterial growth
  • Non-wetting and odor free
  • Clear wall enables visualization of fluid flow
  • Connects easily to ReSeal™ Sanitary Fittings
  • Contains no chemicals listed in California's Prop 65
  • Contains no BPA or phthalates
  • Used to transport non-solvent based chemicals
  • Cosmetic production
  • Food and dairy processing
  • Beverage dispensing
  • Water purification systems
  • Printing equipment
  • Coolant lines
  • Chemical transfer
  • Instrumentation and air supply
Compliance
Meets FDA, 3-A and NSF criteria.
Specifications
Specifications
50 in.
Clear
Product Suggestions
Product Suggestions