Learn More
Saint-Gobain Tygon S3™ E-3603 Flexible Tubings, Unipacs
Formulated for resistance to flex-fatigue and abrasion
Supplier:  Saint-Gobain ACF06036
Encompass
Catalog No. 14171279
Only null left
Description
Description
  • Bio-based formulation and contain no BPA or phthalates
  • Provide high chemical resistance
  • Non-oxidizing and non-contaminating
  • Smooth, polished inner wall helps prevent buildup to facilitate cleaning
  • Coils are marked at 30.4cm (1 ft.) intervals for easy measuring
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Extra-heavy walls withstand full vacuum at room temperature (29.9 in. [759mm] of mercury at 23 °C [73 °F] and up to 27 in. [686mm] of mercury at 60°C [140°F])
  • Sterilized by conventional autoclave methods (steam 30 min. at 15psi, 121°C) and Ethylene Oxide
  • Instrumentation connection, vent, drain and other general laboratory applications
  • Food and beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Condensers
  • Incubators
  • Desiccators
  • Gas lines
  • Drain lines
Compliance
FDA, NSF, 3-A, meets USP Class VI, Japan Food Sanitation Law no. 370/1959, REACH, 1935/2004/EC and 10/2011/EU.
Specifications
Specifications
1/2 in.
10 ft.
10/Pk.
5/8 in.
3.048 m
Clear
Product Suggestions
Product Suggestions