Learn More
Saint-Gobain Tygon S3™ Clear Tubing, Formula B-44-4X, Non-DEHP
Specially designed to ensure bacterial-free fluid path for food processing applications
Supplier:  Saint-Gobain AAA00010
Encompass
Catalog No. 14171145
Only null left
Description
Description
  • Made with phthalate-free, non-DEHP flexible plastic
  • Smooth and non-porous texture prevent from particle entrapment to avoid bacterial growth
  • Good resistance to harsh alkaline cleaners and remain unaffected by common sanitizers
  • Compatible with food that contain high oil
  • Lightweight and easy to handle with minimum requirements of couplings and fittings
  • Available in large bore sizes up to 6 in. inside diameter
  • Provides clear visual inspection and flow control
  • Reinforced tubing available for elevated pressure
Aseptic filling, Condiment dispensing, Dairy processing, Vitamin and flavor concentrate system, Soft-serve dispensing
Compliance
FDA, NSF, 3-A, Japan Food Sanitation Law no. 370/1959, REACH, 1935/2004/EC and 10/2011/EU
Product Suggestions
Product Suggestions