Learn More
Saint-Gobain Tygon™ R-3400 UV Resistant Tubing
For virtually any permanent or temporary chemical transfer application
Supplier: Saint-Gobain AAE00017
Description
- Protects light-sensitive fluids
- Wide range of chemical resistance
- Resistant to ozone
- Resistant to burn
- Low compression set resists permanent deformation
- Black
- Available in clear formulation for fluid flow monitoring
- Acid and caustic transfer
- Corrosive diffusible gases
- Fertilizer and pesticide dispensing
- Electrical insulation lines
- Ink and adhesive dispensing
- General maintenance
Specifications
| 1/4 in. | |
| Tygon | |
| 9.65 mm | |
| Black |
| 6.35 mm | |
| 3/8 in. | |
| 50 ft. |