Learn More
Saint-Gobain Norwell™ Mix-and-Match Standard Bottom with Poppet
Catalog No. 1543751F
Encompass
Change view
Click to view available options
Material:
PTFE, FEP
Catalog No. Material
1543751F PTFE, FEP
1 options
Catalog No. 1543751F Supplier Saint-Gobain Supplier No. D8653214
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Round thread design. Precision ground ball stays confined to the bottom section, preventing sample loss. Special seat prevents leakage and premature opening. Chafe guard reduces wear and abrasion.
Specifications
Product Type Bailer Bottom Cap
Material PTFE, FEP
Style Ball
Diameter (English) Outer 1.87 in.
Diameter (Metric) Outer 4.7 cm