Learn More
Saint-Gobain Norwell™ Mix-and-Match Bailer Extension Coupling DFS Item
Ideal for dedicated or portable applications
Supplier:  Saint-Gobain D8653013
Catalog No. 15-437-51C
Only null left
Description
Description
Allow you to choose the right combination of components to adapt to changing sampling needs and provide a flexible, cost-effective system for monitoring groundwater contamination near landfills and other pollution sources. Precision-machined, interchangeable parts allow quick reassembly after cleaning.
  • For joining bailer bodies to increase bailer capacity
  • With round thread design for easy cleaning and assembly
  • Chafe guard protects bailer body
  • Made of 100% virgin PTFE and FEP
  • Minimize the need for retesting
  • Chemically inert and free of fillers and plasticizers
  • Nonleaching surface resists corrosion and minimizes sample bias
Order Info
Components can be ordered individually, in various combinations, or as part of fully assembled bailer
Specifications
Specifications
Extension Coupling
Product Suggestions
Product Suggestions