missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Norwell™ Mix-and-Match Bailer Bottom Emptying Check Assembly

Catalog No. 1543751G
Encompass
Change view
Click to view available options
Material:
PTFE, FEP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
1543751G PTFE, FEP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1543751G Supplier Saint-Gobain Supplier No. D8653022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for dedicated or portable applications

Allow you to choose the right combination of components to adapt to changing sampling needs and provide a flexible, cost-effective system for monitoring groundwater contamination near landfills and other pollution sources. Precision-machined, interchangeable parts allow quick reassembly after cleaning.

  • Check valve opens when plunger contacts sample collection vessel
  • Contents are emptied with minimal agitation
  • For rapid purging of wells
  • Made of 100% virgin PTFE and FEP
  • Minimize the need for retesting
  • Chemically inert and free of fillers and plasticizers
  • Nonleaching surface resists corrosion and minimizes sample bias

Order Info

Components can be ordered individually, in various combinations, or as part of fully assembled bailer

Specifications

Product Type Bottom Emptying Check Assembly
Material PTFE, FEP
Style Poppet
Diameter (English) Outer 1.87 in.
Diameter (Metric) Outer 4.7 cm