Learn More
Saint-Gobain Fluoroelastomer Fluran™ Tubing
For use in severe environments where other tubings fail—such as peristaltic pump tubing for extremely corrosive materials, solvent recovery systems, dry cleaning fluid lines, and process monitoring equipment.
$3,544.68
Specifications
Material Fluran
Diameter (English) Outer 1/16 in.
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Price Quantity & Availability  
141708C
Encompass
View Documents Promotion Details
Saint-Gobain
AGN00022
5/16 in. 1/16 in.
Case for $3,544.68
Only null left
Description
Description
  • Black, opaque fluoroelastomer tubing (Fluran formula F-5500-A)
  • Resists corrosive chemicals, oils, fuels, solvents, and aliphatic and aromatic hydrocarbons
  • Withstands effects of ozone, sunlight, and weathering
  • Use at temperatures from -40° to +204°C (-40° to +400°F)
  • Durometer hardness: Shore A, 65
  • Not for use with foods, beverages, or drugs
Specifications
Specifications
Fluran
1/16 in.