Learn More
Saint-Gobain Chemfluor™ Fluoropolymer Tubing
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
Supplier:  Saint-Gobain TSPF35050006250
Encompass
Catalog No. 1416917J
Only null left
Description
Description
  • Contains no additives or plasticizers
  • Inert to virtually all chemicals
  • Nonprotein binding
Order Info
In packs of 50 ft. (15m)
Compliance
Complies with FDA criteria
Specifications
Specifications
Complies with FDA criteria
9.5 mm
1/2 in.
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
3/8 in.
PFA
12.7 mm
Product Suggestions
Product Suggestions