missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ SAHARA S21SS Stainless Steel Heated Bath
Description
- Bath volume: 7-17L
- Mateial: Stainless steel
- Ambient +13 to 200°C
Specifications
Specifications
| Capacity (Metric) | 21 L |
| Material | Stainless Steel |