missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SafTest™ Test Kit

Catalog No. MP7KTTG2000
Change view
Click to view available options
Target Molecule:
Alkenal
Free Fatty Acid
Malonaldehyde
Percent Fat
Peroxide
Total Glycerin
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target Molecule
MP7KTTG2000 Total Glycerin
MP7KTAK2000 Alkenal
MP7KTFA2000 Free Fatty Acid
MP7KTMA2000 Malonaldehyde
MP7KTPF2000 Percent Fat
MP7KTPR2000 Peroxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MP7KTTG2000 Supplier MP Biomedicals™ Supplier No. 07KTTG2000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Reagent A, Reagent B, Reagent C, Calibrators

SafTest™ Test Kit measures percentage analytes in given samples. Choose from 6 different kits for use with SafTest instrument and consumable platforms and detection kits.

MP Biomedicals' Test Kits contain all reagents and calibrators necessary to run samples. A control is also included in each Test Kit. Each kit will allow for testing of approximately 100 samples. Sample dilution will likely be required for appropriate analysis.

  • Choose from 6 different kits
  • For use with SafTest instrument and consumable platforms
  • Results in as little as 30 minutes
  • Detection method: Bichromatic Photometer
  • Test Kits contain all reagents and calibrators necessary to run samples.
  • A control is also included in each Test Kit.
  • Each kit will allow for testing of approximately 100 samples.
  • Sample dilution will likely be required for appropriate analysis.
    • Alkenal Test Kit
      • Research Area: Lipid degradation and oxidation
      • Contains all reagents, calibrators
    • Free Fatty Acid Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Calibrators, Control Analyte(s)
      • Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.1 - 2.01% free fatty acid
    • Malonaldehyde Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Calibrators, Control
      • Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.00 to 0.32 mg/kg of sample
    • Percent Fat Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Calibrators, Control Analytes(s)
      • Material Required: SafTest Instrument Platform
      • Quantitation Range: 0.06 to 0
    • Peroxide Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Reagent C, Calibrators, Control Analyte(s)
      • Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.00 to 0.200 MEQ/kg of sample
    • Total Glycerin Test Kit
      • Research Area: Biodiesel
      • Components: Reagent A, Reagent B, Calibrators, Control
      • Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.008 to 0.06% of sample

Specifications

For Use With (Application) Oil/Fat Degradation, Biodiesel Testing
Detection Method Bichromatic Photometer
No. of Reactions 100 Tests
Target Molecule Total Glycerin
Quantity 1 Ea.
Storage Requirements 2°C to 8°C

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.