missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ SafTest™ Test Kit
Description
Includes
Reagent A, Reagent B, Reagent C, Calibrators
MP Biomedicals' Test Kits contain all reagents and calibrators necessary to run samples. A control is also included in each Test Kit. Each kit will allow for testing of approximately 100 samples. Sample dilution will likely be required for appropriate analysis.
- Choose from 6 different kits
- For use with SafTest instrument and consumable platforms
- Results in as little as 30 minutes
- Detection method: Bichromatic Photometer
- Test Kits contain all reagents and calibrators necessary to run samples.
- A control is also included in each Test Kit.
- Each kit will allow for testing of approximately 100 samples.
- Sample dilution will likely be required for appropriate analysis.
- Alkenal Test Kit
- Research Area: Lipid degradation and oxidation
- Contains all reagents, calibrators
- Free Fatty Acid Test Kit
- Research Area: Lipid degradation and oxidation
- Components: Reagent A, Reagent B, Calibrators, Control Analyte(s)
- Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
- Quantitation Range: 0.1 - 2.01% free fatty acid
- Malonaldehyde Test Kit
- Research Area: Lipid degradation and oxidation
- Components: Reagent A, Reagent B, Calibrators, Control
- Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
- Quantitation Range: 0.00 to 0.32 mg/kg of sample
- Percent Fat Test Kit
- Research Area: Lipid degradation and oxidation
- Components: Reagent A, Reagent B, Calibrators, Control Analytes(s)
- Material Required: SafTest Instrument Platform
- Quantitation Range: 0.06 to 0
- Peroxide Test Kit
- Research Area: Lipid degradation and oxidation
- Components: Reagent A, Reagent B, Reagent C, Calibrators, Control Analyte(s)
- Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
- Quantitation Range: 0.00 to 0.200 MEQ/kg of sample
- Total Glycerin Test Kit
- Research Area: Biodiesel
- Components: Reagent A, Reagent B, Calibrators, Control
- Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
- Quantitation Range: 0.008 to 0.06% of sample
Specifications
Specifications
| For Use With (Application) | Percent Fat Testing |
| Target Molecule | Percent Fat |
| Detection Method | Bichromatic Photometer |
| No. of Reactions | 100 Tests |
| Quantity | 1 Ea. |
| Storage Requirements | 2°C to 8°C |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.