missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SafTest™ Sample Consumables Platform

Catalog No. MP07SC2000
Change view
Click to view available options
Sample Type:
Solid Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
MP07SC2000 Solid Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP07SC2000 Supplier MP Biomedicals™ Supplier No. 07SC2000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Printer paper, membranes, precision tissue wipers, solid glass beads, 15mL conical tubes, 10 to 100μL pipette tips, 200 to 1000μL pipette tips, 12mm culture tubes, 12mm culture tube caps, sharpie marke

SafTest™ Sample Consumables Platform contains all consumables necessary to run alkenal, peroxide tests.

  • Consumable platform for conducting alkenal, peroxide testing
  • Contains all of consumables necessary to run solid, liquid samples
  • Research area: Lipid degradation and oxidation
  • Detection method: Bichromatic photometer
  • Application area: Oil/fat degradation

Specifications

For Use With (Application) Peroxide, Alkenal Testing
Sample Type Solid Samples
Quantity 1 Ea.
Storage Requirements Room temperature