missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SafTest™ Preparation Reagent

Catalog No. MP7PREP2000
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP7PREP2000 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP7PREP2000 Supplier MP Biomedicals™ Supplier No. 07PREP2000

missing translation for 'validMainframeMsg'

Contains replacement SafTest™ Preparation Reagent to run all Saftest™ test kits.

  • For use with SafTest analyzer
  • Used with all six SafTest test kits
  • Appropriate for all testing endpoints
  • Research area: Lipid degradation and oxidation
  • Detection method: Bichromatic photometer
  • Application area: Oil/fat degradation

Specifications

For Use With (Application) Oil/Fat Degradation
Detection Method Bichromatic Photometer
Quantity 1 Ea.
Storage Requirements 2°C to 8°C