missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SafTest™ Dispenser Set

Catalog No. MP7DSPR2000
Change view
Click to view available options
For Use With (Application):
Alkenal Testing
Free Fatty Acid Testing
Malonaldehyde Testing
Percent Fat Testing
Peroxide Testing
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
MP7DSPR2000 Peroxide Testing
MP7DSAK2000 Alkenal Testing
MP7DSFA2000 Free Fatty Acid Testing
MP7DSPF2000 Percent Fat Testing
MP7DSMA2000 Malonaldehyde Testing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MP7DSPR2000 Supplier MP Biomedicals™ Supplier No. 07DSPR2000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

One dispenser for each reagent (A, B, and preparation Reagent) and one reagent holding rack

Dispenser Set is an accessory for SafTest™ test kits.

  • Dispenser set for each chosen endpoint
  • Allows for accurate testing, every time
  • For use with SafTest™ kits

Specifications

For Use With (Application) Peroxide Testing
Storage Requirements Room temperature