missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Safranin O, 95%, pure, high purity biological stain

Safranine O, C20H19ClN4, CAS Number-477-73-6, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride, Basic Red 2, C.I. 50240, 100g, 95%, Brown or Brown-Red to Green, 94% min., 350.84, Pure, Warning, GHS H Statement: Causes serious eye irritation. Causes skin irritation. | CAS: 477-73-6 | C20H19ClN4 | 350.85 g/mol

Supplier:  Thermo Scientific Chemicals 146641000

 View more versions of this product

Catalog No. AC146641000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Safranine O
477-73-6
95%
MFCD00011759
QRYAEWIQIBAZOJ-UHFFFAOYSA-N
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
350.85
10% max.
Pure
Brown or Brown/Red to Green
Powder
Pure
94% min.
C20H19ClN4
Basic Red 2, C.I. 50240, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride
Authentic
3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride
2723800
350.84
Glass bottle
100 g

Chemical Identifiers

477-73-6
350.85
QRYAEWIQIBAZOJ-UHFFFAOYSA-N
2723800
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
C20H19ClN4
MFCD00011759
Basic Red 2, C.I. 50240, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride
3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only