missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Safranin O, 85%, pure, certified

CAS: 477-73-6 | C20H19ClN4 | 350.85 g/mol

$115.39

Chemical Identifiers

CAS 477-73-6
Molecular Formula C20H19ClN4
Molecular Weight (g/mol) 350.85
MDL Number MFCD00011759
InChI Key QRYAEWIQIBAZOJ-UHFFFAOYSA-N
Synonym Basic Red 2, C.I. 50240, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride
PubChem CID 2723800
IUPAC Name 3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride
SMILES [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC419210250
View Documents Promotion Details
Thermo Scientific Chemicals
419210250
25 g Glass bottle
Each for $115.39
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

477-73-6
350.85
QRYAEWIQIBAZOJ-UHFFFAOYSA-N
2723800
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
C20H19ClN4
MFCD00011759
Basic Red 2, C.I. 50240, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride
3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride

Specifications

477-73-6
80% min.
C20H19ClN4
Basic Red 2, C.I. 50240, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride
2723800
350.84
Brown to Green
Safranine O, Pure
1.1 to 1.32
85%
MFCD00011759
QRYAEWIQIBAZOJ-UHFFFAOYSA-N
Authentic
350.85
530 to 534nm
Pure
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

RUO – Research Use Only