missing translation for 'onlineSavingsMsg'
Learn More

Safranin, 1% (w/v) Alcoholic, Certified, LabChem™

Supplier:  LabChem LC223601

Catalog No. LC223601


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Safranin
89.1,4.95,4.95,1
-115°C
78°C
Poly Bottle
500 mL
Soluble in water
Certified
1% (w/v) Alcoholic
67-63-0,67-56-1,477-73-6
Red
13°C
C20H19ClN4
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
350.85
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions