missing translation for 'onlineSavingsMsg'
Learn More

Safinamide Free Acid, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023031
Change view
Click to view available options
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111023031 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023031 Supplier Merck Emd Millipore Supplier No. PHR238020MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

Quantity 20 mg
Molecular Formula C17H18FNO3
Linear Formula C17H18FNO3
Formula Weight 303.33
Grade Certified Reference Material