missing translation for 'onlineSavingsMsg'
Learn More

Safinamide Defluor-Derivative, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023032
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023032 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023032 Supplier Merck Emd Millipore Supplier No. PHR238120MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

Flash Point Not Applicable
Molecular Formula C17H20N2O2
Linear Formula C17H20N2O2
Quantity 20 mg
Molecular Weight (g/mol) 284.35
Formula Weight 284.35
Grade Certified Reference Material
Physical Form Powder