missing translation for 'onlineSavingsMsg'
Learn More

Safflower Oil, USP, Spectrum™ Chemical

Catalog No. p-7158146
Change view
Click to view available options
Quantity:
20 L
Packaging:
Steel Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606135 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606135 Supplier Spectrum Chemical Mfg Cor Supplier No. S100420LT
Only null left

S1004, 8001-23-8

Spectrum™ Chemical Safflower Oil, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8001-23-8

Specifications

CAS Max % 1
Packaging Steel Can
Quantity 20 L
Grade USP