Learn More
Medicom™ SafeWear™ Hipster Jacket™, Black
Catalog No. p-7238930
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19910771 Small
19910772 Medium
19910773 Large
19910774 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19910771 Supplier Medicom™ Supplier No. 8118A
Only null left
The SafeWear™ Hipster Jacket™, Black is premium protective apparel made from top-quality materials and designed to provide reliable barrier protection without sacrificing comfort or style.
  • Available in a wide range of styles, colours and sizes
  • Comply with recommended OSHA best practices
  • Made from high-quality 45 g/m2 SMS fabric
  • Detailed finishing, 3-pocket design and front snap closure
  • Each bag contains 10+2 units, for a total of 12 vs. 10 per package from the competition
  • Fluid-resistant fabric meets AAMI PB70 Level 2 standards
  • Fluid-resistant fabric meets AAMI PB70 Level 2 standards
Specifications
Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Color Black
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Latex Free Yes