missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ SafeWear™ High Performance Lab Coats, Tropical Teal

Single-use procedure SMS lab coats are designed to protect health professionals and patients from cross contamination.

$221.03 - $221.03

Specifications

Product Type General Purpose Lab Coat
Gender Unisex
Material SMS Polypropylene
Closure Type Snaps
Cuff Style Knit
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19910841
View Documents
Medicom™
8117A
Small N/A
Only null left
 
19910842
View Documents
Medicom™
8117B
Medium N/A
Only null left
 
19910843
View Documents
Medicom™
8117C
Large N/A
Only null left
 
19910844
View Documents
Medicom™
8117D
X-Large N/A
Only null left
 
Description

Description

  • High quality ultra-soft & light weight SMS fabric (Weight 45 g/m2 for Jacket & Lab Coat)
  • Fluid-resistant material meets AAMI Level 2
  • Designed to fit properly with knitted cuffs & collar
  • High-quality front snap closure buttons
  • Available in a wide range of sizes and in fashionable colors
  • 12 units in every pack
Specifications

Specifications

General Purpose Lab Coat
SMS Polypropylene
Knit
Non-sterile
3
Yes
AAMI, PB70 Level 2, FDA Class 1
Not Autoclavable
Unisex
Snaps
Single-use
Knee-length
Knit
8117
Matching Sewing Thread Color
12 Coats/Pk., 5 Packs/Cs.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'