missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Safety Signs - Multiple Messages
Description
Keep employees safe from harm by effectively communicating hazardous situations to others
- Inform all personnel of a material that is caustic and prompt them to wear PPE.
- Clearly mark empty oxygen cylinders in the workplace to prevent confusion.
- Prevent the mixing of full and empty oxygen cylinders by posting this sign.
- Prevent dangerous situations by requiring personnel to wear fall protection.
Specifications
Specifications
| Material | Vinyl |
| Height (English) | 7 in. |
| Width (English) | 10 in. |
| Displayed Text | Oxygen Full Cylinders |
| Product Type | Safety Sign |
| Color | Green on White |