Learn More
Accuform™ Safety Sign, DANGER LOCK OUT EQUIPMENT BEFORE ENTERING
Catalog No. 19111144
Change view
Click to view available options
Material:
Vinyl
Catalog No. Material
19111144 Vinyl
1 options
Catalog No. 19111144 Supplier Accuform™ Supplier No. MLKT106VS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage. They comply in accordance with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Printed matte surface has permanent acrylic adhesive back
  • Flexible vinyl can be applied to curved or uneven surfaces
  • Moisture resistant for a various light-duty indoor and sheltered outdoor marking needs
  • Square corners and scored back liner for quick peel-off and application
  • 7 x 10, Adhesive Vinyl
Specifications
Material Vinyl
Size (English) 7 x 10 in.
Mounting Type Self-adhesive
Shape Rectangular
Languages English
Displayed Text DANGER LOCK OUT EQUIPMENT BEFORE ENTERING
Color Blue