Learn More
Accuform™ Safety Sign - ATTENTION VACCINE FREEZER NO FOOD OR DRINKS, Aluminum
Accuform™ vaccination signs communicate health and safety policies, reinforce procedures, and identify locations
Supplier:  Accuform™ MBDX915VA
 View more versions of this product
Catalog No. 19-119-131
Only null left
Explore available promotions
Description
Description
  • Thick, rigid aluminum has strength of metal without rust
  • Printed surface has clear UV protective overcoat
  • Withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance
  • Rounded corners and 0.1875 in. mounting holes for installation
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Specifications
    Specifications
    Corner Holes
    Rectangular
    English
    ATTENTION VACCINE FREEZER NO FOOD OR DRINKS DO NOT UNPLUG
    Product Suggestions
    Product Suggestions