Learn More
Sol-Care™ Safety Lancets
Description
- The needle is safely concealed in the lancet body before and after use
- Easy twist-off cap removal
- Comfortable finger recess design
- Small contact face for precise puncturing
Specifications
Specifications
| Sterility | Sterile |
| Material | Polypropylene |
| For Use With (Equipment) | Blood Collection Tube Holder |
| Length (Metric) | 1.6 mm |
| Product Type | Lancet |
| Needle Gauge | 30 G |