missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Safety Information Center: 2-BINDER RTK COMPL CENTER (W/O BINDERS)

Catalog No. 19103248
Change view
Click to view available options
Width (Metric):
438.15 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
19103248 438.15 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19103248 Supplier Brady™ Supplier No. 106346

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Safety information centers provide a centralized location for important safety information and makes the information accessible to employees.

Helps prevent costly mistakes by providing potentially life-saving information to employees

Specifications

Format Wall Mounted
Product Type Information Center
Width (Metric) 438.15 mm
Quantity Kit
Languages English
For Use With (Application) Hazard Communication