missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Safety Bottle Carrier

Catalog No. 16985674
Change view
Click to view available options
Color:
Black
Blue
Green
Red
Size:
1.5 L
4.5 L
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
16985674 Green 1.5 L
16985676 Black 1.5 L
16985675 Blue 1.5 L
16985677 Red 1.5 L
16985679 Blue 4.5 L
16985681 Red 4.5 L
16985680 Black 4.5 L
16985678 Green 4.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 16985674 Supplier Fisherbrand™ Supplier No. 120918F2

The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents in your lab.

  • Robust Design
  • Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
  • Handle can be pressed down to either side for easier loading and unloading
  • Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
  • Carriers come in four colors with designated labeling areas for easy identification
  • Made of chemical resistant thermoplastic elastomer - PP blend

Specifications

Description Safety Bottle Carrier, 1.5 L, Green
Autoclavable Not Autoclavable
Color Green
Holds Bottles
Material Polypropylene
Positions 1
Shape Cylindrical
Stackable No
Sterility Non-sterile
For Use With (Application) Transportation of bottled regeants
For Use With (Equipment) 1.5 L bottles
Size 1.5 L
Temperature (Metric) Range -20 to 60°C
Length (Metric) 11.4 cm
Diameter (Metric) Opening 11.4 cm
Weight (Metric) .25 kg
Includes 1 Safety Bottle Carrier vessel and attachable handle
Product Type Bottle Holder
Diameter (Metric) Inner 11.4 cm
Shelves None
Height (Metric) 16.5 cm (25.4 cm Height, Handle Extended)
Array 1 x 1
Width (Metric) 11.4 cm
Show More Show Less