missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ SafeTouch™ EverStrong™ Vinyl Medical Examination Gloves

Catalog No. 361004634
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
361004634 Small
361004635 Medium
361004636 Large
296358595 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 361004634 Supplier Medicom™ Supplier No. 1129A
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SafeTouch™ EverStrong™ Vinyl Medical Examination Glove offers optimal tactile sensitivity.

  • Heavier weight, highly resistant glove made from latex-free PVC (vinyl).
  • Smooth texture provides optimal tactile sensitivity.
  • 1000/Cs.

Specifications

Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Clear
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Vinyl
Powdered Powder Free
Size Small
Sterility Non-sterile
FDA 510 (k) Yes
Glove Cuff Style Beaded
Show More Show Less