missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ SafeTouch™ Connect™ Latex Medical Examination Gloves

Catalog No. 361004629
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
361004629 X-Small
361004630 Small
361004631 Medium
361004632 Large
361004633 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 361004629 Supplier Medicom™ Supplier No. 1124A
Only null left

SafeTouch™ Connect™ Latex Medical Examination Gloves - Low protein content.

  • Heavier weight, highly resistant, low protein content latex glove.
  • Fully textured for optimal grip.
  • 1000/Cs.

Specifications

Chlorinated No
Accelerator Free No
Applications Healthcare
AQL 1.5
Color Cream
Disposable Single-use
Exterior Finish Fully Textured
Latex Free No
Material Latex
Powdered Powder Free
Size X-Small
Sterility Non-sterile
FDA 510 (k) No
Glove Cuff Style Beaded
Show More Show Less