missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Safetec™ Spill Leader Kit™
Description
Includes
- One 5 oz. bottle of Red-Z (for solidifying and deodorizing biohazardous spills)
- One 5 oz. bottle of Green-Z
- One 5 oz. bottle of Acid Lock solidifier (for encapsulating and neutralizing acids)
- One 5 oz. bottle of Alky solidifier (for encapsulating and neutralizing alkali spills)
- Six 9 x 12 in. zip-pack bags
- Three pairs of nitrile gloves
- Two hazardous waste D.O.T. labels
- Two nonhazardous D.O.T. labels
- Six pick-up scoops with scraper
- One 2 oz. bottle Petro Lock absorbent
- Instructions and MSDS information
Designed for employee protection while handling, cleaning up, or disposing of hazardous liquids. The solid solutions neutralize, stabilize and solidify a variety of aqueous spills.
Specifications
Specifications
| Type | Spill Leader Kit |
| Includes | 3 pairs nitrile gloves, red Z™ solidifier (5 oz. bottle), Green Z™ solidifier (5 oz. bottle), Acid Lock™ solidifier (5 oz. bottle), Alky™ solidifier (5oz. bottle), petro lock absorben |