Learn More
FEATHER Safeshield Scalpel
FEATHER Safeshield scalpel features a clear protective shield that can be retracted or advanced multiple times to cover the blade. Multiple sizes are available with color-coded button on the protective shield to assist in rapid blade size identification.
Supplier: FEATHER SS22
Description
- Made in Japan
- Indispensable in emergencies
- Safety shield can be advanced to cover the blade and retracted multiple times
- Color-coded button on the protective shield for the blade identification
- Individually packed
- Single use only
Chemical Identifiers
| Stainless Steel | |
| Sterile | |
| 10/Pk. |
| Single-use | |
| ISO13485:2016 |
Specifications
| Stainless Steel | |
| Single-use | |
| ISO13485:2016 |
| # 22 | |
| Sterile | |
| 10/Pk. |