missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ SaferGrip™ Mid-Arm Length Cryogenic Gloves

Catalog No. 17355074
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17355074 X-Large
17355071 Small
17355072 Medium
17355073 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17355074 Supplier National Safety Apparel™ Supplier No. G99CRSGPXLMA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gloves offer thermal protection against liquid nitrogen splashes and other cold temperature tasks. SaferGrip™ silicone-coated palm enhances grip and durability.

  • Gloves offer protection from ultra-cold environments in a wide variety of applications
  • 100% waterproof liner
  • Teflon™ Laminated Nylon with 3M™ Scotchlite™ Thinsulate™ liner
  • SaferGrip™ silicone coated material on the palm for enhanced grip and durability
  • Waterproof, breathable membrane for additional splash protection
  • Made in the USA

Specifications

Certifications/Compliance (Europe) EN 511 Contact Cold Performance Level 3, EN 511 Convective Cold Performance Level 3, EN 388 Abrasion Level 4
Chlorinated No
Coated Area Palm
Description Mid-Arm Length
Coating Material Silicone
Color Blue
Exterior Finish Smooth
Hand Specific
Material Nylon
Product Type Cryogenic Gloves
Size X-Large
For Use With (Application) Cryogenic Liquids/Gases
Glove Cuff Style Slip-on
Liner Material 3M™ Thinsulate™
Lining Lined
Series G99CRSGP_ _MA
Length (English) 15 in.
Length (Metric) 38 cm
Product Line SaferGrip Cryogenic Gloves
Show More Show Less