Learn More
Medicom™ SafeMask™ SofSkin Procedure Mask
Catalog No. 17467289
Change view
Click to view available options
Model:
SofSkin ClearFit
SofSkin Fit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
17467289 SofSkin ClearFit
17467288 SofSkin Fit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17467289 Supplier Medicom™ Supplier No. 220014
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SafeMask™ SofSkin skin-friendly procedure ear loop mask with adjustable chin fit for a personalized fit.
  • SofSkin procedure ear loop mask with adjustable chin strip.
  • Skin-friendly multi-layered shingle-pleat mask.
  • Chin piece for personalized comfort.
  • Latex-Free.
  • Procedure masks are intended to be worn by personnel during procedures to protect both the patient and personnel from transfer of microorganisms, body fluids and particulate material transfer.
  • ASTM F2100 standard of conformance.
  • SofSkin ClearFit includes a fog-free strip.
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color White
Fastening Type Earloops
Material Polypropylene
Size One Size
Sterility Non-sterile
Style Pleated
Certifications/Compliance ASTM F2100
Disposable Single-use
Includes Fog Free Strip
Latex Free Yes
Model SofSkin ClearFit
Show More Show Less