Learn More
Medicom™ SafeMask™ Premier Elite Face Masks
Description
- High-barrier mask (ASTM Level 3) for examining or treating high-risk patients and for procedures involving high levels of spray, splatter, moisture and airborne particles
- Smooth, stretchy, latex-free, ultrasonically sealed earloops prevent irritation and allow easy removal with a lower risk of self-contamination
- Adjustable nosepiece provides a comfortable, personalized fit
- Shingle pleat design prevents fluid pooling and cross-contamination
- Non-woven layers will not lint, tear or shred
Specifications
Specifications
| Product Type | Medical/Procedural Face Mask |
| ASTM Level | ASTM Level 3 |
| Color | Lavender |
| Fastening Type | Earloops |
| Sterility | Non-sterile |
| Style | Pleated |
| Disposable | Single-use |
| Latex Free | Yes |