Learn More
Medicom™ SafeMask™ Curve Procedure Mask
Catalog No. 17467290
Change view
Click to view available options
Color:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17467290 White
17467291 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17467290 Supplier Medicom™ Supplier No. 220114
Only null left
SafeMask™ Curve procedure earloop mask provides fuller coverage with a non-pleated design, 3D air pocket which creates a contoured fit that enhances comfort and breathability.
  • SafeMask™ Curve designed to provide protection and comfort for professionals and other users to reduce wearer exposure to and/or transmission of pathogenic particles.
  • Non-pleated mask design includes three folded flaps of ultrasonically sealed, multi-layered filtration material that allows for a contoured fit with a larger '3D' air pocket for greater breathability.
  • The latex-free, fluid-resistant mask includes an adjustable nosepiece and elastic ear loops.
  • Procedure masks are intended to be worn by personnel during procedures to protect both the patient and personnel from transfer of microorganisms, body fluids and particulate material transfer.
  • ASTM F2100 standard of conformance.
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color White
Fastening Type Earloops
Material Polypropylene
Size One Size
Sterility Non-sterile
Style Pleated
Certifications/Compliance ASTM F2100
Disposable Single-use
Latex Free Yes
Show More Show Less