missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ SafeBasics True Fit™ Thin Nitrile Medical Examination Gloves with Textured Fingertips

Catalog No. 296364414
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296364414 X-Small
296364415 Small
296364416 Medium
296364417 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 296364414 Supplier Medicom™ Supplier No. MED1186 A

Medicom™ SafeBasics True Fit™ Thin Nitrile Medical Examination Gloves with Textured Fingertips for optimal tactile sensitivity.

  • Thin Nitrile Medical Examination Gloves
  • Textured Fingertips
  • Pink
  • Optimal tactile sensitivity
  • Chemo tested
  • 3000/Case

Specifications

Chemotherapeutic Rated Yes
Chlorinated No
Accelerator Free No
Applications Healthcare
AQL 1.5
Color Pink
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Small
Sterility Non-sterile
FDA 510 (k) Yes
Glove Cuff Style Beaded
Show More Show Less