Learn More
Medicom™ SafeBasics™ Shoe Covers
Footwear protection with fluid repellant
Supplier: Medicom™ MED8006
Description
- Slip-on
- Sewn seams provide increased tear strength
- Elastic opening for easy on-and-off convenience
- Non-conductive
- Non-Skid and regular sole options
- 100 Shoe Covers/Box, 3 Boxes/Case, 300 Shoe Covers/Case
Specifications
| X-Large | |
| Blue | |
| Non-sterile | |
| Shoe Covers intended to be worn by personnel during various procedures to protect patient/materials and the personnel from transfer of microorganisms, particulates, etc. |
| Non-woven Spun bonded Polypropylene | |
| Single-use | |
| Non-skid | |
| 300/ case |