missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medicom™ SafeBasics™ Beard Cover
Description
- Comfortable face protection
- Elastic edge and headband loop for optimal fit
- Ideal for use in dental, industrial, laboratory, research, and medical industries
Specifications
Specifications
| Size | Universal |
| Color | White |
| Disposable | Single-use |
| Latex Free | Yes |
| For Use With (Application) | Dental, Industrial, Laboratory, Research, Medical |
| Product Type | Cleanroom Beard Cover |