missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Safe Seal System Body Bag Single Facility Pack

Catalog No. 22445886
Change view
Click to view available options
Dimensions (L x W):
150 ft. x 36 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22445886 150 ft. x 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22445886 Supplier Mopec™ Supplier No. BE163

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Safe Seal System of this Single Facility Body Bag Pack is a military tested, heavy duty, hermetically sealable Bio-Safety Level 4 (BSL-4) Containment body bag system. It is easy to use, as the body is placed in the flexible material, then heat-sealed with the Safe Seal Thermosealer to provide complete and secure containment of infectious human or animal remains.

The Safe Seal System was designed to help keep workers and survivors safe after a mass fatality event or any high-risk biohazard exposure. The Facility Pack contains the following:

  • 1 Roll of Mopec™ Safe Seal
  • 1 Heat Sealer
  • 1 Vertical Stand
  • 2 Alignment Clips
  • 1 Razor Knife
  • 1 Extension Cord

Specifications

Closure Type Safe Seal
Color Gray
Material Plastic
Thickness (English) 8 mil
Dimensions (L x W) 150 ft. x 36 in.
Length (English) 150 ft.
Length (English) Document Pouch 45.72 m
Width (English) 36 in.
Product Type Body Bag
Capacity (English) >200 lb.
Length (Metric) 45.72 m
Width (Metric) 91.44 cm
Show More Show Less