missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mopec™ Safe Seal System Body Bag Mass Fatality Pack
Description
The Safe Seal System was designed to help keep workers and survivors safe after a mass fatality event or any high-risk biohazard exposure. The Mass Fatality Pack can handle approximately 750 decedents and contains the following:
- Export Certified Storage Crate
- 30 Rolls of Mopec Safe Seal
- 3 Heat Sealers
- 3 Horizontal Dispensing Racks
- 6 Alignment Clips
- 3 Razor Knives
- 1 Box of 500 Bio Labels
- 1 Box of 500 Adhesive Pouches
Specifications
Specifications
| Closure Type | Safe Seal |
| Color | Gray |
| Material | Plastic |
| Thickness (English) | 8 mil |
| Dimensions (L x W) | 150 ft. x 36 in. |
| Length (English) | 150 ft. |
| Width (English) | 36 in. |
| Product Type | Body Bag |
| Capacity (English) | >200 lb. |
| Length (Metric) | 45.72 m |
| Show More |