missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ Safe Aspiration Station US Plug

Catalog No. F110745G
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F110745G 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F110745G Supplier Gilson™ Supplier No. F110745

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Gilson Safe Aspiration Station US Plug is a stand-alone motorized vacuum system for quick and easy aspiration of biological liquid waste disposal.

  • Liquids are quickly and safely aspirated and collected in a 4 L polypropylene bottle.
  • The station offers adjustable aspiration speed and handles, ideal for ELISA tests, cell culture, and any other contaminated or hazardous liquid aspiration and disposal.

Specifications

Product Type Vacuum
Angle 45°
Autoclavable Autoclavable
Certifications/Compliance CAN/CSA-C22.2 (1010-1)
Coefficient of Variation None
Color Blue
Channels 1
Material PTFE, Polypropylene
Volume (Metric) 4 L
For Use With (Application) Cleaning up laboratory stations
For Use With (Equipment) Stand-alone equipment
Includes 1x Station base with a 4L PP Bottle, 1x Standard tubings set (2 silicone tubes + 1 membrane filter), 1x lever sensor cable, 1x bottle carrying handle, 1x Hand operator, 1x Single-channel plastic tip adaptor with ejector, AC adapter
Quantity 1 Ea.
Warranty 12 Months
Dimensions (L x W x H) 178 X 229 X 457 mm
Length (Metric) 178 mm
Width (English) 9 in
Width (Metric) 229 mm
Height (English) 18 in
Height (Metric) 457 mm
Weight (English) 7.5 lb
Weight (Metric) 3.4 mm
Amperage 15 A
No. per Pack 1
Show More Show Less