missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAF Fixative, Analytical Reagent, MilliporeSigma™
Specifications
Specifications
| Physical Form | Liquid |
| Chemical Name or Material | SAF Fixative |
| Name Note | Analytical Reagent, with Triton (Acetate Buffered Formalin) |
| Color | Colorless |
| Quantity | 200 L |